Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A101372-100MG
A101372-250MG
103795-936EA
128
USD
103795-936
103821-622
76970-160
76970-162
Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride 97%
Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₂·ClH MW: 193.671 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD17171210 CAS Number: 726139-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...