Ethyl-2-(pyrimidin-2-yl)acetate 97%

Supplier: Ambeed
Warning

A178916-250MG A178916-1G A178916-25G
103807-386EA 18 USD
103807-386 103845-724 76784-520 77083-168 77301-588
Ethyl-2-(pyrimidin-2-yl)acetate 97%
Ethyl-2-(pyrimidin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
Boiling Pt: 238 °C (760 mm Hg)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD09953517
CAS Number: 63155-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR