Ethyl-2-(piperidin-4-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(piperidin-4-yl)acetate

A160929-250MG A160929-1G
103805-434EA 40 USD
103805-434 103843-672 77150-880 77319-638
Ethyl-2-(piperidin-4-yl)acetate 97%
Ethyl-2-(piperidin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 232.8 °C (760 mm Hg)
Flash Pt: 94.6 °C
Storage Temperature: Freezer
MDL Number: MFCD02183575
CAS Number: 59184-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR