Ethyl 2-(piperidin-3-yl)acetate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl piperidin-3-ylacetate hydrochloride
A251106-250MG
A251106-1G
A251106-5G
103811-962EA
54
USD
103811-962
103851-804
76801-562
77304-550
77304-552
Ethyl 2-(piperidin-3-yl)acetate hydrochloride 95%
Ethyl 2-(piperidin-3-yl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₈ClNO₂ MW: 207.7 g/mol Melting Pt: 146 °C Storage Temperature: Ambient |
MDL Number:
MFCD08059314 CAS Number: 16780-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...