Ethyl-2-(oxetan-3-ylidene)acetate 97%

Supplier: Ambeed
Warning

A300844-100MG A300844-25G A300844-250MG
103814-288EA 16 USD
103814-288 103835-438 76875-664 76875-666 77883-826
Ethyl-2-(oxetan-3-ylidene)acetate 97%
Ethyl-2-(oxetan-3-ylidene)acetate

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
Boiling Pt: 207.1 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Freezer
MDL Number: MFCD12755197
CAS Number: 922500-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR