Ethyl-4-hydroxy-2-(methylthio)pyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(methylthio)-6-oxo-1,6-dihydropyrimidine-5-carboxylate, Ethyl 1,4-dihydro-2-(methylthio)-4-oxopyrimidine-5-carboxylate

A136541-100G A136541-10G A136541-500G A136541-1G A136541-25G A136541-5G
103802-316EA 5 USD
103802-316 76767-544 76767-546 76767-548 77298-854 77315-226 77315-228
Ethyl-4-hydroxy-2-(methylthio)pyrimidine-5-carboxylate 95%
Ethyl-4-hydroxy-2-(methylthio)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
Boiling Pt: 340.4 °C (760 mm Hg)
Melting Pt: 136…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023234
CAS Number: 53554-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR