Ethyl-2-(methylamino)benzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-(methylamino)benzoate
A269234-5G
A269234-250MG
103812-954EA
101
USD
103812-954
103852-860
77092-620
77672-388
Ethyl-2-(methylamino)benzoate 95%
Ethyl-2-(methylamino)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂ Boiling Pt: 226.2 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD11174593 CAS Number: 35472-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...