Ethyl 2-(imidazo[1,2-a]pyridin-3-yl)acetate 97%

Supplier: Ambeed
Warning

A199799-100MG A199799-250MG A199799-1G
103817-136EA 18 USD
103817-136 103847-762 77012-600 77012-602 77302-632
Ethyl 2-(imidazo[1,2-a]pyridin-3-yl)acetate 97%
Ethyl 2-(imidazo[1,2-a]pyridin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13183145
CAS Number: 101820-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR