Ethyl-2-acetyl-3-ethoxyacrylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(ethoxymethylene)acetoacetate, Ethyl 2-(ethoxymethylene)-3-oxobutanoate

A194206-100G A194206-10G A194206-1G A194206-25G
103816-632EA 9 USD
103816-632 103847-244 76974-988 76974-990 77085-164 77085-166
Ethyl-2-acetyl-3-ethoxyacrylate 95%
Ethyl-2-acetyl-3-ethoxyacrylate

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
Boiling Pt: 268.8 °C (760 mm Hg)
Flash Pt: 113.7 °C
Storage Temperature: Ambient
MDL Number: MFCD09959866
CAS Number: 3788-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR