Triethyl-2-phosphonopropionate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate

A104496-100G A104496-10G A104496-25G A104496-1G A104496-5G A104496-500G
103796-724EA 5 USD
103796-724 103822-438 76780-730 76780-732 76780-734 77309-304 77309-306 77565-824
Triethyl-2-phosphonopropionate 98%
Triethyl-2-phosphonopropionate

Precaución: For research use only.

Formula: C₉H₁₉O₅P
MW: 238.22 g/mol
Boiling Pt: 275.5 °C (760 mm Hg)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00009159
CAS Number: 3699-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR