Ethyl (Benzyloxy)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(benzyloxy)acetate, (Benzyloxy)acetic acid Ethyl ester

A110259-100G A110259-10G A110259-25G A110259-1G A110259-5G
103797-644EA 5 USD
103797-644 103823-452 76869-682 76869-684 76869-686 77310-248 77310-250
Ethyl (Benzyloxy)acetate 97%
Ethyl (Benzyloxy)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 284.6 °C (760 mm Hg)
Flash Pt: 98.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06204614
CAS Number: 32122-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR