N-Benzylglycine ethyl ester 96%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-(benzylamino)acetate

A204904-100G A204904-10G A204904-1000G A204904-25G
103809-404EA 250 USD
103809-404 76814-424 76814-426 77302-916 77302-918
N-Benzylglycine ethyl ester 96%
N-Benzylglycine ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 277 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009174
CAS Number: 6436-90-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR