Ethyl 2-(2-Benzothiazolyl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Benzothiazoleacetic acid Ethyl ester, Ethyl 2-Benzothiazoleacetate, 2-(2-Benzothiazolyl)acetic acid Ethyl ester

A318154-100G A318154-10G A318154-250MG A318154-1G A318154-5G A318154-25G
103825-162EA 18 USD
103825-162 103836-094 76772-886 76772-888 76772-890 76838-944 76838-946 77306-172
Ethyl 2-(2-Benzothiazolyl)acetate 97%
Ethyl 2-(2-Benzothiazolyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂S
MW: 221.27 g/mol
Boiling Pt: 331 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00546397
CAS Number: 29182-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR