Ethyl-2-(aminomethyl)isonicotinate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-(aminomethyl)isonicotinate hydrochloride
A155771-1G
A155771-5G
A155771-100MG
103804-798EA
66
USD
103804-798
103843-000
77318-732
77318-734
77667-198
Ethyl-2-(aminomethyl)isonicotinate hydrochloride 97%
Ethyl-2-(aminomethyl)isonicotinate hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₃ClN₂O₂ MW: 216.6700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09788627 CAS Number: 1189983-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...