Ethyl 2-(6-chloropyridin-2-yl)acetate 97%
Supplier: Ambeed
|
Warning
|
A663381-100MG
A663381-250MG
A663381-1G
A663381-5G
103840-454EA
325
USD
103840-454
76819-806
76819-808
76844-644
76844-646
Ethyl 2-(6-chloropyridin-2-yl)acetate 97%
Ethyl 2-(6-chloropyridin-2-yl)acetate
Precaución: for Research Use Only
Formula:
C₉H₁₀ClNO₂ Boiling Pt: 268.7 °C (760 mm Hg) Flash Pt: 116.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD21604163 CAS Number: 174666-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...