Ethyl 2-(5-bromo-3-nitropyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A251533-100MG A251533-250MG A251533-1G
103811-986EA 40 USD
103811-986 103851-828 77165-988 77165-990 77304-560
Ethyl 2-(5-bromo-3-nitropyridin-2-yl)acetate 95%
Ethyl 2-(5-bromo-3-nitropyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₄
MW: 289.08 g/mol
Boiling Pt: 334.7±37.0 °C (760 mm Hg)
Flash Pt: 156.2 °C
Storage Temperature: Ambient
MDL Number: MFCD16876027
CAS Number: 1211540-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR