Ethyl-2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

A164535-100G A164535-10G A164535-1G A164535-25G A164535-5G
103805-876EA 9 USD
103805-876 103844-132 77028-758 77028-760 77300-720 77300-722 77300-724
Ethyl-2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 98%
Ethyl-2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃S
MW: 263.31 g/mol
Boiling Pt: 426.8 °C (760 mm Hg)
Melting Pt: 180 °C
Flash Pt: 211.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03700424
CAS Number: 161797-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR