Ethyl 2-(4-formylphenyl)acetate ≥95%

Supplier: Ambeed
Warning

A156159-100MG A156159-250MG A156159-1G
103804-840EA 152 USD
103804-840 103843-044 77162-950 77162-952 77300-160
Ethyl 2-(4-formylphenyl)acetate ≥95%
Ethyl 2-(4-formylphenyl)acetate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15523550
CAS Number: 43153-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR