Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate 97%

Supplier: Ambeed
Danger

A256060-100MG A256060-250MG A256060-1G A256060-5G A256060-10G
103812-234EA 30 USD
103812-234 103852-094 76827-060 76827-062 77091-568 77091-570 77683-779
Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate 97%
Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate

Precaución: For research use only.

Formula: C₉H₁₁ClN₂O₂S
MW: 246.714 g/mol
Boiling Pt: 345.8 °C (760 mm Hg)
Flash Pt: 162.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977460
CAS Number: 61727-34-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR