Ethyl 2-(4-bromopyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A706728-100MG A706728-250MG A706728-1G A706728-5G
103840-674EA 494 USD
103840-674 76820-072 76820-074 76844-886 76844-888
Ethyl 2-(4-bromopyridin-2-yl)acetate 95%
Ethyl 2-(4-bromopyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566271
CAS Number: 1060814-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR