Ethyl-2-(4-bromophenyl)thiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-bromophenyl)thiazole-4-carboxylate

A130058-250MG A130058-1G
103801-370EA 103 USD
103801-370 76983-652 77313-802
Ethyl-2-(4-bromophenyl)thiazole-4-carboxylate 95%
Ethyl-2-(4-bromophenyl)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂S
MW: 312.19 g/mol
Boiling Pt: 404.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06738340
CAS Number: 885278-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR