2-(Ethoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A214744-10G A214744-250MG A214744-1G A214744-5G A214744-25G
103818-432EA 19 USD
103818-432 103849-136 76785-780 76785-782 77087-664 77087-666 77303-376
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester 97%
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
Boiling Pt: 378 °C (760 mm Hg)
Melting Pt: 60…65 °C
Flash Pt: 182.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03453660
CAS Number: 269409-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR