Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-(trifluoromethyl)phenyl)acetate, Ethyl 4-(Trifluoromethyl)phenylacetate, 4-(Trifluoromethyl)phenylacetic acid ethyl ester

A227851-100G A227851-10G A227851-250MG A227851-1G A227851-5G
103819-342EA 7 USD
103819-342 103850-250 76801-008 76801-010 76801-012 77089-036 77089-038
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 98%
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₂
Boiling Pt: 239.6 °C (760 mm Hg)
Flash Pt: 73.897 °C
Storage Temperature: Ambient
MDL Number: MFCD07779351
CAS Number: 721-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR