Ethyl (3-Hydroxyphenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(3-hydroxyphenyl)acetate

A333382-1G A333382-5G A333382-10G A333382-250MG A333382-25G
103825-638EA 6 USD
103825-638 103836-604 76839-608 76839-610 77015-446 77015-448 77015-450
Ethyl (3-Hydroxyphenyl)acetate 98%
Ethyl (3-Hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 292.5 °C (760 mm Hg)
Flash Pt: 123.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07369387
CAS Number: 22446-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR