Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(3-formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

A313303-1G A313303-5G A313303-250MG
103825-002EA 9 USD
103825-002 103835-914 76838-730 76838-732 77032-650
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 98%
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₄S
MW: 291.32 g/mol
Boiling Pt: 446.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13194810
CAS Number: 161798-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR