Ethyl 2-(3-cyano-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A100507-100G A100507-10G A100507-25G A100507-500G A100507-1G A100507-5G
103795-490EA 13 USD
103795-490 103821-184 76869-090 76869-092 76869-094 76869-096 77296-734 77296-736
Ethyl 2-(3-cyano-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate 97%
Ethyl 2-(3-cyano-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₃S
MW: 288.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144699
CAS Number: 161798-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR