(3-Chlorophenyl)acetic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl (3-chlorophenyl)acetate, Ethyl 2-(3-chlorophenyl)acetate

A544448-100G A544448-25G A544448-1G A544448-5G
103828-654EA 9 USD
103828-654 103839-776 76805-400 76843-840 77307-782 77307-784
(3-Chlorophenyl)acetic acid ethyl ester 98%
(3-Chlorophenyl)acetic acid ethyl ester

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198.65 g/mol
Boiling Pt: 258.431 °C (760 mm Hg)
Flash Pt: 118.312 °C
Storage Temperature: Ambient
MDL Number: MFCD07787498
CAS Number: 14062-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR