Ethyl-2-(3-bromopyridin-4-yl)acetate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-(3-bromopyridin-4-yl)acetate

A612657-100MG A612657-250MG A612657-1G A612657-5G
103840-176EA 274 USD
103840-176 77035-190 77035-192 77307-944 77307-946
Ethyl-2-(3-bromopyridin-4-yl)acetate 97%
Ethyl-2-(3-bromopyridin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
Boiling Pt: 296.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD17011773
CAS Number: 51054-99-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR