Ethyl 2-(3-bromo-2-methylphenyl)acetate 97%

Supplier: Ambeed
Warning

A100576-100MG A100576-250MG A100576-1G
103795-518EA 35 USD
103795-518 103821-206 77233-034 77233-036 77282-512
Ethyl 2-(3-bromo-2-methylphenyl)acetate 97%
Ethyl 2-(3-bromo-2-methylphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
Boiling Pt: 305.4±27.0 °C (760 mm Hg)
Flash Pt: 138.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18395994
CAS Number: 1261862-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR