Ethyl 2-nitrophenylacetate 97%

Supplier: Ambeed
Warning

A166244-10G A166244-250MG A166244-25G A166244-5G
103806-032EA 36 USD
103806-032 103844-292 77273-554 77273-556 77320-536 77320-538
Ethyl 2-nitrophenylacetate 97%
Ethyl 2-nitrophenylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
Boiling Pt: 298.9 °C (760 mm Hg)
Flash Pt: 128.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00092226
CAS Number: 31912-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR