Ethyl 2-(2-chlorophenyl)acetate 97%

Supplier: Ambeed
Warning

A152755-100G A152755-10G A152755-25G
103842-600EA 5 USD
103842-600 77028-234 77028-236 77318-230 77318-232
Ethyl 2-(2-chlorophenyl)acetate 97%
Ethyl 2-(2-chlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Boiling Pt: 258.4 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09264019
CAS Number: 40061-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR