Ethyl 2-(2-(methylamino)pyrimidin-4-yl)-1H-indole-5-carboxylate 96%

Supplier: Ambeed
Warning

A102047-100MG A102047-250MG A102047-1G
103796-266EA 325 USD
103796-266 76869-180 76869-182 77282-896
Ethyl 2-(2-(methylamino)pyrimidin-4-yl)-1H-indole-5-carboxylate 96%
Ethyl 2-(2-(methylamino)pyrimidin-4-yl)-1H-indole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₄O₂
MW: 296.32 g/mol
Boiling Pt: 552.6±58.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD17018712
CAS Number: 916486-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR