Ethyl 2-(1-methylguanidino)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A104706-100MG A104706-250MG A104706-1G
103796-762EA 125 USD
103796-762 103822-484 76687-192 76687-194 77309-348
Ethyl 2-(1-methylguanidino)acetate hydrochloride 95%
Ethyl 2-(1-methylguanidino)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN₃O₂
MW: 195.65 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704792
CAS Number: 15366-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR