ethyl-2-((tert-butoxycarbonyl)amino)acetate 97%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)glycine ethyl ester, ethyl 2-((tert-Butoxycarbonyl)amino)acetate, Boc-Gly-OEt, N-Boc-glycine ethyl ester

A258398-10G A258398-500G A258398-25G A258398-5G
103852-246EA 5 USD
103852-246 76786-918 76786-920 77304-730 77304-732
ethyl-2-((tert-butoxycarbonyl)amino)acetate 97%
ethyl-2-((tert-butoxycarbonyl)amino)acetate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.2 g/mol
Flash Pt: 125.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663720
CAS Number: 14719-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR