Ethyl-1-methylpiperidine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 1-methylpiperidine-4-carboxylate
A182323-25G
A182323-5G
A182323-100G
A182323-10G
A182323-1G
103807-696EA
7
USD
103807-696
103846-066
77083-606
77083-608
77163-916
77163-918
77301-776
Ethyl-1-methylpiperidine-4-carboxylate 97%
Ethyl-1-methylpiperidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₂ MW: 171.2400 g/mol Boiling Pt: 211 °C (760 mm Hg) Flash Pt: 87.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130015 CAS Number: 24252-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...