Ethyl 1-methylcyclopropane-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Ethoxycarbonyl)-1-methylcyclopropane, Ethyl 1-methylcyclopropanecarboxylate

A746622-250MG A746622-1G A746622-5G
103840-834EA 96 USD
103840-834 77118-146 77308-246 77308-248
Ethyl 1-methylcyclopropane-1-carboxylate 95%
Ethyl 1-methylcyclopropane-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.169 g/mol
Boiling Pt: 136 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00001284
CAS Number: 71441-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR