Ethyl 1-methyl-4-oxocyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A298163-100MG A298163-250MG A298163-1G
103814-176EA 91 USD
103814-176 103835-320 76816-702 76816-704 77305-736
Ethyl 1-methyl-4-oxocyclohexanecarboxylate 97%
Ethyl 1-methyl-4-oxocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₃
MW: 184.23 g/mol
Boiling Pt: 252.9±33.0 °C (760 mm Hg)
Flash Pt: 104.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17018639
CAS Number: 147905-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR