Ethyl-1-methyl-4-nitroimidazole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

A147589-250MG A147589-1G A147589-25G A147589-5G
103803-806EA 32 USD
103803-806 103841-912 76783-234 77317-298 77317-300 77317-302
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate 97%
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
Boiling Pt: 350.7 °C (760 mm Hg)
Flash Pt: 165.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03789110
CAS Number: 109012-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR