Ethyl-1-methyl-1H-imidazole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylimidazole-2-carboxylic acid ethyl ester, Ethyl 1-methylimidazole-2-carboxylate, Ethyl 1-methyl-1H-imidazole-2-carboxylate, Ethyl 1-Methylimidazole-2-carboxylate

A212275-10G A212275-1G A212275-25G A212275-5G
103818-182EA 7 USD
103818-182 103848-882 76800-676 77087-342 77087-344 77087-346
Ethyl-1-methyl-1H-imidazole-2-carboxylate 98%
Ethyl-1-methyl-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
Boiling Pt: 254.8 °C (760 mm Hg)
Melting Pt: 29…31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683326
CAS Number: 30148-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR