Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A158868-1G A158868-25G A158868-5G A158868-10G
103805-180EA 13 USD
103805-180 103843-394 77300-374 77319-248 77319-250 77671-250
Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate 95%
Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
Boiling Pt: 355.9 °C (760 mm Hg)
Melting Pt: 179…181 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD11053752
CAS Number: 17288-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR