Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-azaindole-2-carboxylate, Ethyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate

A240232-1G A240232-100MG
103811-344EA 15 USD
103811-344 103851-142 77090-296 77667-972
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 97%
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2000 g/mol
Boiling Pt: 355.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09998715
CAS Number: 221675-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR