1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-pyrazolo[4,3-b]pyridine-3-carboxylate

A233419-100MG A233419-1G
103810-888EA 179 USD
103810-888 103850-648 76987-454 77089-606
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
Boiling Pt: 369 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16658534
CAS Number: 1234616-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR