Ethyl indole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Indole-3-carboxylic acid ethyl ester, Ethyl 1H-indole-3-carboxylate

A343591-100G A343591-1G
103825-888EA 5 USD
103825-888 103836-866 76773-202 76839-910
Ethyl indole-3-carboxylate 97%
Ethyl indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 120…124 °C
Flash Pt: 160.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00228454
CAS Number: 776-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR