Imidazole-2-carboxylic acid ethyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate
A149401-100G
A149401-10G
A149401-250MG
A149401-500G
A149401-1G
A149401-25G
A149401-5G
103804-040EA
6
USD
103804-040
103842-158
77162-700
77162-702
77162-704
77162-706
77317-652
77317-654
77317-656
Imidazole-2-carboxylic acid ethyl ester 98%
Imidazole-2-carboxylic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Boiling Pt: 269.5 °C (760 mm Hg) Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426031 CAS Number: 33543-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...