Ethyl 1-cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A119500-250MG
A119500-1G
A119500-5G
103799-776EA
32
USD
103799-776
76870-154
77297-692
77297-694
Ethyl 1-cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate 97%
Ethyl 1-cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₃F₂NO₃ MW: 293.27 g/mol Boiling Pt: 419.5 °C (760 mm Hg) Melting Pt: 227 °C Flash Pt: 207.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646378 CAS Number: 98349-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...