Ethyl-1-cyanocyclobutanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-cyanocyclobutanecarboxylate

A416279-1G A416279-25G A416279-5G
103827-312EA 7 USD
103827-312 103838-358 76841-938 76841-940 76841-942
Ethyl-1-cyanocyclobutanecarboxylate 98%
Ethyl-1-cyanocyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 242.8 °C (760 mm Hg)
Flash Pt: 106.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04114273
CAS Number: 28246-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR