Ethyl 1-bromocyclopropanecarboxylate 97%

Supplier: Ambeed
Warning

A213750-100MG A213750-10G A213750-250MG A213750-1G A213750-5G
103818-328EA 22 USD
103818-328 103849-038 76814-656 76814-658 76814-660 77087-546 77087-548
Ethyl 1-bromocyclopropanecarboxylate 97%
Ethyl 1-bromocyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₆H₉BrO₂
Boiling Pt: 60 °C (10 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01314227
CAS Number: 89544-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR