Ethyl-1-benzylpyrrolidine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-benzylpyrrolidine-3-carboxylate

A754288-100MG A754288-10G A754288-25G A754288-1G A754288-5G
103840-862EA 18 USD
103840-862 76791-374 76791-376 76791-378 76845-096 76845-098
Ethyl-1-benzylpyrrolidine-3-carboxylate 97%
Ethyl-1-benzylpyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.31 g/mol
Boiling Pt: 308.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04039852
CAS Number: 5747-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR