Ethyl 1-benzyl-4-oxopyrrolidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A126363-5G A126363-100G A126363-10G
103800-800EA 9 USD
103800-800 77313-014 77571-568 77666-798
Ethyl 1-benzyl-4-oxopyrrolidine-3-carboxylate 95%
Ethyl 1-benzyl-4-oxopyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.28 g/mol
Boiling Pt: 358.1±42.0 °C (760 mm Hg)
Flash Pt: 170.4 °C
Storage Temperature: Freezer
MDL Number: MFCD09998956
CAS Number: 1027-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR