Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A211648-100G
A211648-10G
A211648-500G
A211648-25G
A211648-5G
103848-826EA
8
USD
103848-826
76785-686
76785-688
76785-690
77087-248
77303-208
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₃·HCl MW: 297.780 g/mol Melting Pt: 175 °C Flash Pt: 183.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012800 CAS Number: 1454-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...